Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 45-4732-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: This eBMG2b antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBMG2b antibody has been tested by flow cytometric analysis of normal human peripheral blood and mouse splenocytes. Use at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2b kappa | |
eBMG2b | |
PerCP-Cyanine5.5 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.