missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa, PE-eFluor 610, clone: eBMG2b, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 61-4732-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15520507

  • £62.06 / 25µg


Estimated Shipment date
from Supplier 04-06-2018
Add to basket

Description

Description

Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin.Applications Reported: Mouse IgG2b K Isotype Control PE-eFluor® 610 has been reported for use in flow cytometric analysis.Applications Tested: Mouse IgG2b K Isotype Control PE-eFluor® 610 has been tested by flow cytometric analysis of normal human peripheral blood cells. Use the isotype control at the same concentration as the experimental antibody.PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2b, K
Control
PE-eFluor 610
Conjugated
Murine
Affinity chromatography
RUO
Isotype Control
Flow Cytometry
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG2, IgG2b, IgG2bk
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.