missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa, PE-eFluor 610, clone: eBMG2b, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 61-4732-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15530507

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin.
Specifications

Specifications

IgG2b, K
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Flow Cytometry
PE-eFluor 610
Conjugated
IgG2b, kappa
RUO
Monoclonal
Videos

For Research Use Only.