missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa, PE-Cyanine5, clone: eBMG2b, Isotype Control, eBioscience™

Mouse Isotype Control

£59.25 - £97.75

Specifications

Antigen Mouse IgG2b kappa
Clone eBMG2b
Target Species Not Applicable
Conjugate PE-Cyanine5
Isotype IgG2b, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15577756
View Documents
Affymetrix eBioscience
15-4732-41
25 Tests
£59.25
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
08-01-2021
Add to basket
 
15587756
View Documents
Affymetrix eBioscience
15-4732-42
100 Tests
£97.75
100 tests
View alternative products
Save Up to 
»
Estimated Shipment
08-01-2021
Add to basket
 
Description

Description

Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin.Applications Reported: This mouse IgG2b isotype control has been reported for use in flow cytometric analysis.Applications Tested: Mouse IgG2b K Isotype Control PE-Cy5 has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mouse IgG2b kappa
Not Applicable
IgG2b, kappa, kappa
Monoclonal
Control
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO
eBMG2b
PE-Cyanine5
Control, Flow Cytometry
Liquid
Isotype Control
Store at 2-8°C. Do not freeze. Light-sensitive material.
SDS

For Research Use Only.