Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), PE, eBioscience™, Invitrogen™
Mouse Isotype Control
£74.25 - £123.00
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Applications | Control, Flow Cytometry |
Conjugate | PE |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15238779
|
Affymetrix eBioscience
12-4732-81 |
50 μg |
£74.25
50µg |
In Stock
Log in to see stock. |
|||||
15248779
|
Affymetrix eBioscience
12-4732-82 |
100 μg |
£123.00
100µg |
In Stock
Log in to see stock. |
|||||
15286568
|
Affymetrix eBioscience
12-4732-42 |
100 Tests |
£92.25
100 tests |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
Description
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: This mouse IgG2b isotype control has been reported for use in flow cytometric analysis. Applications Tested: This Mouse IgG2b K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and can be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2b kappa | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBMG2b | |
PE | |
Mouse | |
Not Applicable | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.