Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 50-4732-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: Mouse IgG2b K Isotype Control eFluor® 660 has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: Mouse IgG2b K Isotype Control eFluor® 660 has been tested by flow cytometric analysis. This should be used at the same concentration as the experimental antibody. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Specifications
Mouse IgG2b kappa | |
eBMG2b | |
eFluor 660 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.