missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 570, eBioscience™, Invitrogen™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 41-4732-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15578936

  • £307.00 / 100µg
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: This eBMG2b antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, immunohistochemical staining, and immunocytochemistry. Applications Tested: This eBMG2b antibody has been tested by immunocytochemistry on fixed and permeabilized cells. Use isotype control at same concentration as experimental antibody. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm

Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).
Specifications

Specifications

Mouse IgG2b kappa
eBMG2b
eFluor 570
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
100 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.