missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa, Functional Grade, clone: eBMG2b, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 16-4732-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258627

  • £112.49 / 500µg


EU Stock 8
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin.Applications Reported: This mouse IgG2b isotype control has been reported for use in flow cytometric analysis.Applications Tested: This mouse IgG2b Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

IgG2b κ
eBMG2b
Unconjugated
Purified
IgG2b
RUO
Monoclonal
Flow Cytometry
1mg/mL
aqueous buffer, no sodium azide
Murine
500μg
Store at 2-8°C.
Videos