missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG2a, PE-Cyanine7, clone: m2a-15F8, Secondary Antibody, eBioscience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 25-4210-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15518546

  • £68.27 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.Applications Reported: This m2a-15F8 antibody has been reported for use in flow cytometric analysis.Applications Tested: This m2a-15F8 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2a
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
Affinity chromatography
RUO
Secondary Antibody
Monoclonal
Flow Cytometry
m2a-15F8
PE-Cyanine7
Conjugated
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Secondary
Murine
Videos

For Research Use Only.