missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgG2a, FITC, clone: m2a-15F8, Secondary Antibody, eBioscience™
Rat Monoclonal Secondary Antibody
Brand: Affymetrix eBioscience 11-4210-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.Applications Reported: This m2a-15F8 antibody has been reported for use in immunohistochemical staining and immunocytochemistry.Applications Tested: This m2a-15F8 antibody has been tested by immunocytochemistry on fixed MDCK cells at less than or equal to 20 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
IgG2a | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Rat | |
Affinity chromatography | |
RUO | |
Secondary Antibody | |
Monoclonal |
Immunohistochemistry | |
m2a-15F8 | |
FITC | |
Conjugated | |
IgG1, kappa | |
25μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Secondary | |
Murine |
For Research Use Only.