missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG2a, Biotin, clone: m2a-15F8, Secondary Antibody, eBioscience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 13-4210-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537336

  • £89.95 / 100µg


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody m2a-15F8 recognizes Mouse IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG2a or any rat isotype antibodies.Applications Reported: This m2a-15F8 antibody has been reported for use in immunohistochemical staining and immunocytochemistry.Applications Tested: This m2a-15F8 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue and by immunocytochemistry on fixed MDCK cells. The antibody can be used at less than or equal to 10 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2a
m2a-15F8
Biotin
Conjugated
IgG1
RUO
Primary
Murine
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos