missing translation for 'onlineSavingsMsg'
Learn More

IgG2a Rat anti-Mouse, APC, eBioscience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 17-4210-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15588086

  • £52.50 / 25µg

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Mouse IgG2a
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Rat
Affinity chromatography
RUO
Secondary Antibody
Monoclonal
Flow Cytometry
m2a-15F8
APC
Liquid
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Secondary
Mouse
Videos
Documents

For Research Use Only.