Learn More
Mouse anti-Rat IgG2a, PE-Cyanine7, clone: r2a-21B2, Secondary Antibody, eBioscience™
Mouse Monoclonal Secondary Antibody
Brand:  Affymetrix eBioscience 25-4817-80
Code : NEW
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 15578546
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
The monoclonal antibody r2a-21B2 recognizes Rat IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2a or any mouse isotype antibodies.
Specifications
Specifications
IgG2a
r2a-21B2
PE-Cyanine7
Conjugated
IgG1, kappa
RUO
Primary
Rat
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.