Learn More
Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Secondary Antibody
£48.28 - £139.00
Specifications
Antigen | Rat IgG2a |
---|---|
Clone | r2a-21B2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15509966
|
Affymetrix eBioscience
50-4817-82 |
100 μg |
£139.00
100µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15599956
|
Affymetrix eBioscience
50-4817-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody r2a-21B2 recognizes Rat IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2a or any mouse isotype antibodies. Applications Reported: This r2a-21B2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This r2a-21B2 antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood cells stained with a rat IgG2a primary antibody. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a Monoclonal antibody specifically detects Rat IgG2a in Rat samples. It is validated for Flow Cytometry.Specifications
Rat IgG2a | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 κ | |
Affinity chromatography |
r2a-21B2 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Rat | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.