missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG2a, eFluor 615, clone: r2a-21B2, Secondary Antibody, eBioscience™

Mouse Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 42-4817-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15351110

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The monoclonal antibody r2a-21B2 recognizes Rat IgG2a antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2a or any mouse isotype antibodies.
Specifications

Specifications

IgG2a
r2a-21B2
eFluor 615
Conjugated
IgG1, kappa
RUO
Primary
Rat
Immunohistochemistry (Frozen)
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.