missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2a kappa, PE-Cyanine5.5, clone: eBR2a, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 35-4321-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15301040

  • £49.78 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin.Applications Reported: This eBR2a antibody has been reported for use in flow cytometric analysis.Applications Tested: This rat IgG2a isotype control has been tested by flow cytometric analysis of mouse splenocyte suspension. This should be used at the same concentration as the experimental antibody.Excitation: 488-567 nm; Emission: 690 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Specifications

Specifications

IgG2a, K
Control
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG2, IgG2a, IgG2k
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
eBR2a
PE-Cyanine5.5
Conjugated
Rat
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.