Learn More
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 615, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 42-4321-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG2a Isotype Control eFluor® 615 has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: Rat IgG2a Isotype Control eFluor® 615 has been tested by immunocytochemistry or immunohistochemistry. Use at the same concentration as the experimental antibody. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor® 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75). A standard Alexa Fluor® 594 filter is acceptable. Excitation: 595 nm; Emission: 615 nm. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry.Specifications
Rat IgG2a kappa | |
eBR2a | |
eFluor 615 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a κ |
For Research Use Only.