missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2a kappa, eFluor 570, clone: eBR2a, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 41-4321-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15321070

  • £70.27 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin.Applications Reported: This eBR2a antibody has been reported for use in immunohistochemical and immunocytochemical staining.Applications Tested: This Rat IgG2a Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized C2C12 cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nm
Specifications

Specifications

IgG2a, K
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG2, IgG2a, IgG2k
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Immunohistochemistry
eBR2a
eFluor 570
Conjugated
Rat
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.