Learn More
Rat IgG2a kappa Isotype Control (eBR2a), eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 14-4321-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in immunocytochemistry, immunohistochemistry, flow cytometric analysis, and immunoglobulin ELISA. Applications Tested: This rat IgG2a isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and should be used at the same concentration as the experimental antibody. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.Specifications
Rat IgG2a kappa | |
eBR2a | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Isotype Control (Primary) | |
4° C | |
IgG2a κ |
For Research Use Only.