missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2a kappa, PerCP-eFluor™ 710, clone: eBM2a, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 46-4724-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15371320

  • £148.80 / 100µg


EU Stock 4
Add to basket

Description

Description

Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies.Applications Reported: PerCP-eFluor® 710 Mouse IgG2a, K Isotype Control has been reported for use in flow cytometric analysis.Applications Tested: PerCP-eFluor® 710 Mouse IgG2a Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells.Use at the same concentration as experimental antibody.PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2a κ
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG2, IgG2a, IgG2ak
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
eBM2a
PerCP-eFluor 710
Conjugated
Murine
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.