Learn More
Mouse IgG2a kappa Isotype Control (eBM2a), PE, eBioscience™, Invitrogen™
Mouse Isotype Control
£47.64 - £116.00
Specifications
Antigen | Mouse IgG2a kappa |
---|---|
Clone | eBM2a |
Applications | Control, Flow Cytometry |
Conjugate | PE |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15218779
|
Affymetrix eBioscience
12-4724-81 |
50 μg |
£75.00
50µg |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
15228779
|
Affymetrix eBioscience
12-4724-82 |
100 μg |
£116.00
100µg |
In Stock
Log in to see stock. |
|||||
15556976
|
Affymetrix eBioscience
12-4724-41 |
25 Tests |
£47.64
25 tests |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15182076
|
Affymetrix eBioscience
12-4724-42 |
100 Tests |
£99.00
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
Description
Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: This mouse IgG2a isotype control has been reported for use in cell surface and intracellular flow cytometric analysis. Applications Tested: This Mouse IgG2a K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and can be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2a kappa | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBM2a | |
PE | |
Mouse | |
Not Applicable | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.