missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2a kappa, clone: eBM2a, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 14-4724-85

Code : Z2

Additional Details : Weight : 0.01000kg

Product Code. 15207377

  • £81.84 / 500µg


EU Stock 5
Add to basket

Description

Description

Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies.Applications Reported: This mouse IgG2a isotype control has been reported for use in surface and intracellular flow cytometric analysis, immunocytochemistry, and immunohistochemistry.Applications Tested: This mouse IgG2a isotype control has been tested by flow cytometric analysis of human peripheral blood leukocytes and should be used at at the same concentration as the experimental antibody.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG2a, K
Control
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG2, IgG2a, IgG2ak
IgG2a, kappa
500μg
Store at 2-8°C.
Monoclonal
Flow CytometryþImmunocytochemistryþImmunohistochemistry
eBM2a
Unconjugated
Purified
Murine
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.