Learn More
Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 506, eBioscience™, Invitrogen™
Mouse Isotype Control
£69.50 - £164.00
Specifications
Antigen | Mouse IgG2a kappa |
---|---|
Clone | eBM2a |
Conjugate | eFluor 506 |
Form | Liquid |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15500687
|
Affymetrix eBioscience
69-4724-82 |
100 μg |
£164.00
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15590677
|
Affymetrix eBioscience
69-4724-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This is a monoclonal mouse IgG2a antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: This eBM2a antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBM2a antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2a kappa Control antibody specifically detects Mouse IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2a kappa | |
eFluor 506 | |
Mouse | |
IgG2a κ | |
Affinity chromatography |
eBM2a | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.