missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2a kappa, eFluor 506, clone: eBM2a, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 69-4724-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15590677

  • £65.25 / 25µg


Estimated Shipment
23-11-2020
Add to basket

Description

Description

Description: This is a monoclonal mouse IgG2a antibody. It is used as an isotype control for mouse IgG2a antibodies.Applications Reported: This eBM2a antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBM2a antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody.eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mouse IgG2a kappa
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG2, IgG2a, IgG2ak
IgG2a, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Control, Flow Cytometry
eBM2a
eFluor 506
Liquid
Murine
Affinity chromatography
RUO
Isotype Control
Not Applicable
Videos
Documents

For Research Use Only.