Learn More
Mouse IgG2a kappa Isotype Control (eBM2a), APC, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 17-4724-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: This mouse IgG2a isotype control has been reported for use in flow cytometric analysis. Applications Tested: The APC conjugated mouse IgG2a isotype control is offered in 2 formats: - μg size: has been tested by flow cytometric analysis of normal human peripheral blood cells and can be used at less than or equal to 1 μg/million cells (100 μL blood). It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells and can be used at test size: 5 μL (1 μg)/test per million cells (100 μL blood). Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2a kappa Control antibody specifically detects Mouse IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2a kappa | |
eBM2a | |
APC | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a κ |
For Research Use Only.