Learn More
Mouse IgG2a kappa Isotype Control (eBM2a), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 53-4724-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This is a monoclonal mouse IgG2a, kappa antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: Alexa Fluor® 488 Mouse IgG2a, K Isotype Control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: Alexa Fluor® 488 Mouse IgG2a, K Isotype Control has been tested. This can be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2a kappa Control antibody specifically detects Mouse IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Specifications
Mouse IgG2a kappa | |
eBM2a | |
Alexa Fluor 488 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2a κ |
For Research Use Only.