missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG1, PE-eFluor 610, Clone: M1-14D12, Secondary Antibody, eBiocience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 61-4015-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15580487

  • £45.69 / 25µg

Estimated Shipment date
from Supplier 27-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies.Applications Reported: This M1-14D12 antibody has been reported for use in flow cytometric analysis.Applications Tested: This M1-14D12 antibody has been tested by flow cytometric analysis of staining with a mouse IgG1 primary antibody. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mouse IgG1
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
Affinity chromatography
RUO
Secondary Antibody
Monoclonal
Flow Cytometry
M1-14D12
PE-eFluor 610
Liquid
IgG
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Secondary
Murine
Videos
Documents

For Research Use Only.