missing translation for 'onlineSavingsMsg'
Learn More

IgG1 Rat anti-Mouse, PE-eFluor® 610, eBioscience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 61-4015-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15590487

  • £112.00 / 100µg

Estimated Shipment
01-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Mouse IgG1
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
Affinity chromatography
RUO
Secondary Antibody
Monoclonal
Flow Cytometry
M1-14D12
PE-eFluor™ 610
Liquid
IgG
100 μg
4° C, store in dark, DO NOT FREEZE!
Secondary
Mouse
Videos
Documents

For Research Use Only.