missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG1, HRP, clone: M1-14D12, Secondary Antibody, eBioscience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 18-4015-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15518306

  • £94.99 / 100µg


Estimated Shipment date
from Supplier 27-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies.Applications Reported: This M1-14D12 antibody has been reported for use in western blotting, ELISA, immunohistochemical and immunocytochemical staining.Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry and immunohistochemistry after staining with a primary antibody of the mouse IgG1 isotype. This can be used at less than or equal to 20μg/mL. For western blotting, this can be used at 1μg/mL (1/500 dilution). It is recommended that the antibody be titrated for optimal performance in the assay of interest.
Specifications

Specifications

IgG1
M1-14D12
HRP
Conjugated
IgG
RUO
Primary
Murine
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos