Learn More
Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™, Invitrogen™
Rat Monoclonal Secondary Antibody
Brand: Affymetrix eBioscience 42-4015-80
80.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in immunohistochemical staining and immunocytochemistry. Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry and immunohistochemistry after staining with a primary antibody of the mouse IgG1 isotype. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be titrated for optimal performance in the assay of interest. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor™ 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75). A standard Alexa Fluor™ 594 filter is acceptable. Excitation: 595 nm; Emission: 615 nm. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 Monoclonal antibody specifically detects Mouse IgG1 in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Mouse IgG1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
M1-14D12 | |
eFluor 615 | |
Rat | |
25 μg | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |
For Research Use Only.