missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG1, Biotin, clone: M1-14D12, Secondary Antibody, eBioscience™

Rat Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 13-4015-80

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15389293

  • £37.98 / 25µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy applications. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies.Applications Reported: This M1-14D12 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry on fixed and permeabilized cells using a mouse IgG1 primary antibody. This antibody can be used at less than or equal to 10 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1
M1-14D12
Biotin
Conjugated
IgG
RUO
Primary
Murine
Immunohistochemistry (Frozen)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos