missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG1, PE, clone: R1-12D10, Secondary Antibody, eBioscience™

Mouse Monoclonal Secondary Antibody

Brand:  Affymetrix eBioscience 12-4812-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15536986

  • £43.71 / 25µg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Description

Description

Description: The R1-12D10 monoclonal antibody recognizes rat IgG1 antibodies and can be used as a second step reagent in microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG1.Applications Reported: This R1-12D10 antibody has been reported for use in flow cytometric analysis.Applications Tested: This R1-12D10 antibody has been tested by flow cytometric analysis of mouse splenocytes stained with a rat IgG1 primary antibody. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Murine
Affinity chromatography
RUO
Secondary Antibody
Monoclonal
Flow Cytometry
R1-12D10
PE
Conjugated
IgG
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Secondary
Rat
Videos

For Research Use Only.