missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1, κ, Mouse, Alexa Fluor 647, Clone: MOPC-21, Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Brand: BD Biosciences 565571
Additional Details : Weight : 0.01000kg
Description
IgG1, κ Monoclonal antibody specifically detects IgG1, κ in samples. It is validated for Flow cytometry, Isotype control.Specifications
IgG1, κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Purified |
Flow Cytometry | |
MOPC-21 | |
Alexa Fluor 647 | |
Mouse | |
RUO | |
Primary | |
IgG1 κ |
For Reseach Use Only.