missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG1 kappa Isotype Control (eBRG1), PE-Cyanine5, eBioscience™, Invitrogen™
Rat Isotype Control
£113.00 - £182.00
Specifications
Antigen | Rat IgG1 kappa |
---|---|
Clone | eBRG1 |
Conjugate | PE-Cyanine5 |
Form | Liquid |
Host Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15370510
|
Affymetrix eBioscience
15-4301-82 |
100 μg |
£182.00
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15360510
|
Affymetrix eBioscience
15-4301-81 |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: PE-Cy5 Rat IgG1 Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: This PE-Cy5 Rat IgG1 isotype control antibody should be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG1 is a subclass of immunoglobulin G (IgG) antibodies that are commonly used in research and diagnostics. These antibodies are produced by rats in response to an antigen and can be purified from serum or hybridoma cell culture supernatants. Rat IgG1 is often used as a negative control in experiments, as it does not bind to many mammalian antigens. It is also used in immunohistochemistry, flow cytometry, and western blotting to detect specific antigens. Gene targeting is a powerful tool used to manipulate the rat genome and generate animal models for studying human diseases. With gene targeting, specific genes can be knocked out, inserted, or modified to investigate their role in disease pathogenesis.Specifications
Rat IgG1 kappa | |
PE-Cyanine5 | |
Rat | |
IgG1 κ | |
Affinity chromatography |
eBRG1 | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.