missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa, FITC, clone: eBRG1, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 11-4301-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278209

  • £49.78 / 50µg


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA.Applications Tested: Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1, K
eBRG1
FITC
Conjugated
IgG1
RUO
Monoclonal
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Videos