Learn More
Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 570, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 41-4301-80
98.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This eBRG1 antibody has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: This eBRG1 antibody has been tested by immunohistochemistry on frozen mouse tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry.Specifications
Rat IgG1 kappa | |
eBRG1 | |
eFluor 570 | |
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
For Research Use Only.