Learn More
Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 450, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 48-4301-80
43.83 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG1 Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: eFluor® 450 Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG1 kappa | |
eBRG1 | |
eFluor 450 | |
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
For Research Use Only.