Learn More
Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 450, eBioscience™, Invitrogen™
Rat Isotype Control
£65.75 - £149.00
Specifications
Antigen | Rat IgG1 kappa |
---|---|
Clone | eBRG1 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | eFluor 450 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15321770
|
Affymetrix eBioscience
48-4301-80 |
25 μg |
£65.75
25µg |
Estimated Shipment: 15-04-2024
Log in to see stock. |
|||||
15331770
|
Affymetrix eBioscience
48-4301-82 |
100 μg |
£149.00
100µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG1 Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: eFluor® 450 Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG1 kappa | |
0.2 mg/mL | |
eFluor 450 | |
Rat | |
Not Applicable | |
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBRG1 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.