missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa, Functional Grade, clone: eBRG1, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 14-4301-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15277357

  • £75.09 / 500Āµg


EU Stock 5
Add to basket

Description

Description

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and immunoglobulin ELISA.Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and should be used at the same concentration as the experimental antibody.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1 κ
Control
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
500μg
Store at 2-8°C.
Monoclonal
Flow CytometryĆ¾ImmunocytochemistryĆ¾Immunohistochemistry
eBRG1
Functional Grade
Purified
Rat
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.