missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa, Functional Grade, clone: eBRG1, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 14-4301-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15257357

  • £30.74 / 50µg


Estimated Shipment
27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and immunoglobulin ELISA.Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and should be used at the same concentration as the experimental antibody.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1 κ
eBRG1
Unconjugated
Purified
IgG1
RUO
Monoclonal
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
50μg
Store at 2-8°C.
Videos