missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa, Biotin, clone: eBRG1, Isotype Control, eBioscience™

Rat Isotype Control

Brand:  Affymetrix eBioscience 13-4301-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15223598

  • £37.06 / 50µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: This rat IgG1 isotype control can be used in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA.Applications Tested: This rat IgG1 isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IgG1 κ
Control
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Immunohistochemistry
eBRG1
Biotin
Conjugated
Rat
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.