missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa, PE-eFluor 610, clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Isotype Control

£65.75 - £162.00

Specifications

Antigen Mouse IgG1 kappa
Clone P3.6.2.8.1
Target Species Not Applicable
Conjugate PE-eFluor 610
Isotype IgG1, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15590497 View Documents Affymetrix eBioscience
61-4714-80
25μg £65.75
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-04-2020
Add to basket
 
15500507 View Documents Affymetrix eBioscience
61-4714-82
100μg £162.00
100µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-04-2020
Add to basket
 
Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: This P3.6.2.8.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. Use the isotype control at the same concentration as the experimental antibody.PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mouse IgG1 kappa
Not Applicable
IgG1, kappa, kappa
Monoclonal
Control
PBS with 0.09% sodium azide; pH 7.2
RUO
P3.6.2.8.1
PE-eFluor 610
Control, Flow Cytometry
Liquid
Isotype Control
Store at 2-8°C. Do not freeze. Light-sensitive material.

For Research Use Only.