Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PE-Cyanine5.5, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 35-4714-82
108.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: PE-Cy5.5 Mouse IgG1, K Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of human peripheral blood cells and mouse splenocytes. Excitation: 488-561 nm; Emission: 695 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG1 kappa | |
P3.6.2.8.1 | |
PE-Cyanine5.5 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
For Research Use Only.