missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa, PE-Cyanine5.5, clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 35-4714-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15321040

  • £49.78 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control.Applications Reported: PE-Cy5.5 Mouse IgG1, K Iso Cntrl has been reported for use in flow cytometric analysis.Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of human peripheral blood cells and mouse splenocytes.Excitation: 488-567 nm; Emission: 690 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Specifications

Specifications

IgG1, K
Control
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
P3.6.2.8.1
PE-Cyanine5.5
Conjugated
Murine
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.