Learn More
Mouse IgG1 kappa, PE, clone: P3.6.2.8.1, Isotype Control, eBioscience™
Mouse Isotype Control
Brand: Affymetrix eBioscience 12-4714-42
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control.Applications Reported: PE Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis.Applications Tested: This Mouse IgG1 K Isotype Control is offered in 2 formats: - μg size: has been tested by flow cytometric analysis. Use isotype control at the same concentration as experimental antibody. - test size: has been pre-titrated and tested by flow cytometric analysis. This can be used at test size: 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Mouse IgG1 kappa | |
Control | |
5μl/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
IgG1, IgG1k | |
IgG1, kappa, kappa | |
100 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Monoclonal |
Control, Flow Cytometry | |
P3.6.2.8.1 | |
PE | |
Liquid | |
Murine | |
Affinity chromatography | |
RUO | |
Isotype Control | |
Not Applicable |
For Research Use Only.