missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa, eFluor 570, clone: P3.6.2.8.1, Isotype Control, eBioscience™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 41-4714-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15548936

  • £64.20 / 25Āµg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control immunoglobulin.Applications Reported: This P3.6.2.8.1 antibody has been reported for use in immunohistochemical and immunocytochemical staining.Applications Tested: This Mouse IgG1 K Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized MDCK cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human brain tissue. Use isotype control at the same concentration as experimental antibody.Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nm
Specifications

Specifications

IgG1, K
Control
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG1, IgG1k
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
ImmunocytochemistryĆ¾Immunohistochemistry
P3.6.2.8.1
eFluor 570
Conjugated
Murine
Affinity chromatography
RUO
Isotype Control
Videos

For Research Use Only.