Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Isotype Control
£129.00 - £277.00
Specifications
Antigen | Mouse IgG1 kappa |
---|---|
Clone | P3.6.2.8.1 |
Conjugate | eFluor 570 |
Form | Liquid |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558936
|
Affymetrix eBioscience
41-4714-82 |
100 μg |
£277.00
100µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15548936
|
Affymetrix eBioscience
41-4714-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control immunoglobulin. Applications Reported: This P3.6.2.8.1 antibody has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: This Mouse IgG1 K Isotype Control eFluor® 570 has been tested by immunocytochemistry on fixed and permeabilized MDCK cells and by immunohistochemistry (IHC-P) on formalin-fixed paraffin embedded human brain tissue. Use isotype control at the same concentration as experimental antibody. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Mouse IgG1 kappa | |
eFluor 570 | |
Mouse | |
IgG1 κ | |
Affinity chromatography |
P3.6.2.8.1 | |
Liquid | |
RUO | |
Isotype Control (Primary) |
For Research Use Only.