Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 48-4714-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: Mouse IgG1 K Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: This Mouse IgG1 K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG1 kappa | |
P3.6.2.8.1 | |
eFluor 450 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
For Research Use Only.