Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 14-4714-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: This mouse IgG1 isotype control has been reported for use in surface and intracellular flow cytometric analysis, immunohistochemistry, immunocytochemistry, immunoprecipitation and immunoblotting (WB). Applications Tested: Mouse IgG1 K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG1 kappa | |
P3.6.2.8.1 | |
Unconjugated | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C | |
IgG1 κ |
For Research Use Only.