Learn More
Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), APC, eBioscience™, Invitrogen™
Mouse Isotype Control
£72.75 - £124.00
Specifications
Antigen | Mouse IgG1 kappa |
---|---|
Clone | P3.6.2.8.1 |
Applications | Control, Flow Cytometry |
Conjugate | APC |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15300730
|
Affymetrix eBioscience
17-4714-81 |
50 μg |
£72.75
50µg |
In Stock
Log in to see stock. |
|||||
15310730
|
Affymetrix eBioscience
17-4714-82 |
100 μg |
£116.00
100µg |
In Stock
Log in to see stock. |
|||||
15528096
|
Affymetrix eBioscience
17-4714-41 |
25 Tests |
£74.25
25 tests |
In Stock
Log in to see stock. |
|||||
12642059
|
Affymetrix eBioscience
17-4714-42 |
100 Tests |
£124.00
Each |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: APC Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: The Mouse IgG1 K Isotype Control is offered in 2 formats:- μg size: This can be used at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested by intracellular staining for flow cytometric analysis. This can be used at test size: 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG1 kappa | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
P3.6.2.8.1 | |
APC | |
Mouse | |
Not Applicable | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.